Anti-PAPPA Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: PAPPA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77810-186
- 77810-185
- Antigen Symbol:
- PAPPA
- PAPPA
- Antigen Name:
- Pappalysin-1 protein (95-302AA)
- Pappalysin-1 protein (95-302AA)
- Antigen Synonyms:
- ASBABP2|Aspecific BCL2 ARE binding protein 2|Differentially placenta 1 expressed protein|DIPLA1|EC 3.4.24.79|IGF dependent IGFBP 4 protease|IGF-dependent IGFBP-4 protease|IGFBP 4ase|IGFBP-4ase|IGFBP4ase|Insulin like growth factor dependent IGF binding protein 4 protease|Insulin-like growth factor-dependent IGF-binding protein 4 protease|PAPA|PAPP A|PAPP-A|PAPP1_HUMAN|PAPPA|PAPPA1|Pappalysin 1 precursor|Pappalysin-1|Pregnancy Associated Plasma Protein A|Pregnancy associated plasma protein A pappalysin 1|Pregnancy-associated plasma protein A
- ASBABP2|Aspecific BCL2 ARE binding protein 2|Differentially placenta 1 expressed protein|DIPLA1|EC 3.4.24.79|IGF dependent IGFBP 4 protease|IGF-dependent IGFBP-4 protease|IGFBP 4ase|IGFBP-4ase|IGFBP4ase|Insulin like growth factor dependent IGF binding protein 4 protease|Insulin-like growth factor-dependent IGF-binding protein 4 protease|PAPA|PAPP A|PAPP-A|PAPP1_HUMAN|PAPPA|PAPPA1|Pappalysin 1 precursor|Pappalysin-1|Pregnancy Associated Plasma Protein A|Pregnancy associated plasma protein A pappalysin 1|Pregnancy-associated plasma protein A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13219
- Q13219
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together