Anti-ANGPTL3 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ANGPTL3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77808-902
- 77808-903
- Antigen Symbol:
- ANGPTL3
- ANGPTL3
- Antigen Name:
- Angiopoietin-related protein 3 protein (17-268AA)
- Angiopoietin-related protein 3 protein (17-268AA)
- Antigen Synonyms:
- ANG 5|ANG-5|ANG5|Angiopoietin 5|Angiopoietin like 3|Angiopoietin related protein 3|Angiopoietin-5|Angiopoietin-like protein 3|Angiopoietin-related protein 3|ANGL3_HUMAN|ANGPT5|ANGPTL3|ANL3|FHBL2|OTTHUMP00000010719|UNQ153/PRO179
- ANG 5|ANG-5|ANG5|Angiopoietin 5|Angiopoietin like 3|Angiopoietin related protein 3|Angiopoietin-5|Angiopoietin-like protein 3|Angiopoietin-related protein 3|ANGL3_HUMAN|ANGPT5|ANGPTL3|ANL3|FHBL2|OTTHUMP00000010719|UNQ153/PRO179
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y5C1
- Q9Y5C1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together