Anti-AZI2 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: AZI2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-222
- 77836-223
- Antigen Symbol:
- AZI2
- AZI2
- Antigen Name:
- 5-azacytidine-induced protein 2 protein (258-376AA)
- 5-azacytidine-induced protein 2 protein (258-376AA)
- Antigen Synonyms:
- 5 azacytidine induced protein 2|5-azacytidine-induced protein 2|AA410145|AZ2|AZI2|AZI2_HUMAN|MGC112644|Nak associated protein 1|Nak-associated protein 1|NAP1|NF kappa B activating kinase associated protein 1|NF-kappa-B-activating kinase-associated protein 1|TILP|TILP(S)
- 5 azacytidine induced protein 2|5-azacytidine-induced protein 2|AA410145|AZ2|AZI2|AZI2_HUMAN|MGC112644|Nak associated protein 1|Nak-associated protein 1|NAP1|NF kappa B activating kinase associated protein 1|NF-kappa-B-activating kinase-associated protein 1|TILP|TILP(S)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H6S1
- Q9H6S1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together