Anti-CCDC68 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CCDC68
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-579
- 77840-580
- Antigen Symbol:
- CCDC68
- CCDC68
- Antigen Name:
- Coiled-coil domain-containing protein 68 protein (131-335AA)
- Coiled-coil domain-containing protein 68 protein (131-335AA)
- Antigen Synonyms:
- CCD68_HUMAN| CCDC 68| CCDC68| Coiled coil domain containing 68| Coiled coil domain containing protein 68| Coiled-coil domain-containing protein 68| CTCL associated antigen se57 1| CTCL tumor antigen se57 1| CTCL-associated antigen se57-1| Cutaneous T cell lymphoma associated antigen| Cutaneous T cell lymphoma associated antigen se57 1| Cutaneous T-cell lymphoma-associated antigen se57-1| FLJ25368| SE57 1
- CCD68_HUMAN| CCDC 68| CCDC68| Coiled coil domain containing 68| Coiled coil domain containing protein 68| Coiled-coil domain-containing protein 68| CTCL associated antigen se57 1| CTCL tumor antigen se57 1| CTCL-associated antigen se57-1| Cutaneous T cell lymphoma associated antigen| Cutaneous T cell lymphoma associated antigen se57 1| Cutaneous T-cell lymphoma-associated antigen se57-1| FLJ25368| SE57 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H2F9
- Q9H2F9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together