Anti-SSB Rabbit Recombinant Polyclonal Antibody
Catalog # 77801-088
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Lupus La protein (1-408AA)
- Antigen Symbol:SSB
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P05455
- Antigen Synonyms:Autoantigen La|La |La autoantigen|La autoantigen homolog|La protein|La ribonucleoprotein|La ribonucleoprotein domain family member 3|LA_HUMAN|LARP3|Lupus La antigen|Lupus La protein|Lupus La protein homolog|MGC118101|MGC93380|mRNA for autoantigen|OTTMUSP00000014043|RP23 273G23.1|Sjoegren syndrome type B antigen|Sjogren syndrome antigen B (autoantigen La)|Sjogren syndrome antigen B|SS B|SS-B|SS-B/La protein|SSB
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: SSB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse