Anti-OTUD7B Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: OTUD7B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-533
- 77830-534
- Antigen Symbol:
- OTUD7B
- OTUD7B
- Antigen Name:
- OTU domain-containing protein 7B protein (235-347AA)
- OTU domain-containing protein 7B protein (235-347AA)
- Antigen Synonyms:
- Cellular zinc finger anti NF kappa B protein |Cellular zinc finger anti-NF-kappa-B protein|CEZANNE|HGNC:16683|OTU domain containing protein 7B|OTU domain-containing protein 7B|OTU7B_HUMAN|OTUD7B|ZA20D1|Zinc finger A20 domain containing protein 1|Zinc finger A20 domain-containing protein 1|Zinc finger protein Cezanne
- Cellular zinc finger anti NF kappa B protein |Cellular zinc finger anti-NF-kappa-B protein|CEZANNE|HGNC:16683|OTU domain containing protein 7B|OTU domain-containing protein 7B|OTU7B_HUMAN|OTUD7B|ZA20D1|Zinc finger A20 domain containing protein 1|Zinc finger A20 domain-containing protein 1|Zinc finger protein Cezanne
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6GQQ9
- Q6GQQ9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together