Anti-UBAC2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500
Type:
Antigen: UBAC2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-428
- 77833-429
- Antigen Symbol:
- UBAC2
- UBAC2
- Antigen Name:
- Ubiquitin-associated domain-containing protein 2 protein (1-157AA)
- Ubiquitin-associated domain-containing protein 2 protein (1-157AA)
- Antigen Synonyms:
- UBAC2| PHGDHL1| PSEC0110| Ubiquitin-associated domain-containing protein 2| UBA domain-containing protein 2| Phosphoglycerate dehydrogenase-like protein 1
- UBAC2| PHGDHL1| PSEC0110| Ubiquitin-associated domain-containing protein 2| UBA domain-containing protein 2| Phosphoglycerate dehydrogenase-like protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NBM4
- Q8NBM4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together