Anti-UNC50 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UNC50
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77826-982
- 77826-983
- Antigen Symbol:
- UNC50
- UNC50
- Antigen Name:
- Protein unc-50 homolog protein (15-82AA)
- Protein unc-50 homolog protein (15-82AA)
- Antigen Synonyms:
- UNC50|UNCL|HSD-23|HSD23|Protein unc-50 homolog|Periodontal ligament-specific protein 22|PDLs22|Protein GMH1 homolog|hGMH1|Uncoordinated-like protein
- UNC50|UNCL|HSD-23|HSD23|Protein unc-50 homolog|Periodontal ligament-specific protein 22|PDLs22|Protein GMH1 homolog|hGMH1|Uncoordinated-like protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q53HI1
- Q53HI1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together