Anti-PLA2G10 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PLA2G10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77794-287
- 77794-288
- Antigen Symbol:
- PLA2G10
- PLA2G10
- Antigen Name:
- Group 10 secretory phospholipase A2 protein (43-165AA)
- Group 10 secretory phospholipase A2 protein (43-165AA)
- Antigen Synonyms:
- Group 10 secretory phospholipase A2|Group X secretory phospholipase A2|GX sPLA2|GXPLA2|GXSPLA2|PA2GX_HUMAN|Phosphatidylcholine 2 acylhydrolase GX|Phosphatidylcholine 2-acylhydrolase 10|Phospholipase A2 group X|PLA2G10|SPLA2|sPLA2-X|sPLA2X
- Group 10 secretory phospholipase A2|Group X secretory phospholipase A2|GX sPLA2|GXPLA2|GXSPLA2|PA2GX_HUMAN|Phosphatidylcholine 2 acylhydrolase GX|Phosphatidylcholine 2-acylhydrolase 10|Phospholipase A2 group X|PLA2G10|SPLA2|sPLA2-X|sPLA2X
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15496
- O15496
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together