Anti-DHX36 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: DHX36
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-509
- 77835-510
- Antigen Symbol:
- DHX36
- DHX36
- Antigen Name:
- ATP-dependent RNA helicase DHX36 protein (68-181AA)
- ATP-dependent RNA helicase DHX36 protein (68-181AA)
- Antigen Synonyms:
- DDX36| DEAH box protein 36| Dhx36| DHX36_HUMAN| EC 3.6.1.-| KIAA1488| MLE like protein 1| MLE-like protein 1| MLEL1| Probable ATP dependent RNA helicase DHX36| Probable ATP-dependent RNA helicase DHX36| RHAU| RNA helicase associated with AU rich element ARE | RNA helicase associated with AU-rich element ARE
- DDX36| DEAH box protein 36| Dhx36| DHX36_HUMAN| EC 3.6.1.-| KIAA1488| MLE like protein 1| MLE-like protein 1| MLEL1| Probable ATP dependent RNA helicase DHX36| Probable ATP-dependent RNA helicase DHX36| RHAU| RNA helicase associated with AU rich element ARE | RNA helicase associated with AU-rich element ARE
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H2U1
- Q9H2U1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together