Anti-EHHADH Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: EHHADH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77821-507
- 77821-506
- Antigen Symbol:
- EHHADH
- EHHADH
- Antigen Name:
- Peroxisomal bifunctional enzyme protein (1-723AA)
- Peroxisomal bifunctional enzyme protein (1-723AA)
- Antigen Synonyms:
- 3 hydroxyacyl CoA dehydrogenase|3,2 trans enoyl CoA isomerase|3-hydroxyacyl-CoA dehydrogenase|ECHD|ECHP_HUMAN|Enoyl Coenzyme A|hydratase/3 hydroxyacyl Coenzyme A dehydrogenase|L 3 hydroxyacyl CoA dehydrogenase|L bifunctional protein|peroxisomal|L PBE|LBFP|LBP|MGC120586|MS730|PBE|PBFE|Peroxisomal bifunctional enzyme|Peroxisomal enoyl CoA hydratase
- 3 hydroxyacyl CoA dehydrogenase|3,2 trans enoyl CoA isomerase|3-hydroxyacyl-CoA dehydrogenase|ECHD|ECHP_HUMAN|Enoyl Coenzyme A|hydratase/3 hydroxyacyl Coenzyme A dehydrogenase|L 3 hydroxyacyl CoA dehydrogenase|L bifunctional protein|peroxisomal|L PBE|LBFP|LBP|MGC120586|MS730|PBE|PBFE|Peroxisomal bifunctional enzyme|Peroxisomal enoyl CoA hydratase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q08426
- Q08426
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together