Anti-TSPAN15 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: TSPAN15
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77803-630
- 77811-630
- Antigen Symbol:
- TSPAN15
- TSPAN15
- Antigen Name:
- Tetraspanin-15 protein (115-235AA)
- Tetraspanin-15 protein (115-235AA)
- Antigen Synonyms:
- Tetraspan NET-7|Tetraspanin 15|Tetraspanin-15|Transmembrane 4 superfamily member 15|TSN15_HUMAN|Tspan-15
- Tetraspan NET-7|Tetraspanin 15|Tetraspanin-15|Transmembrane 4 superfamily member 15|TSN15_HUMAN|Tspan-15
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95858
- O95858
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together