Anti-TTC38 Rabbit Recombinant Polyclonal Antibody
Catalog # 77828-163
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Tetratricopeptide repeat protein 38 protein (2-200AA)
- Antigen Symbol:TTC38
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q5R3I4
- Antigen Synonyms:LL22NC03-5H6.5|Tetratricopeptide repeat protein 38|TPR repeat protein 38|TTC38|TTC38_HUMAN
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: TTC38
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse