Anti-TPD52L2 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TPD52L2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-360
- 77803-360
- Antigen Symbol:
- TPD52L2
- TPD52L2
- Antigen Name:
- Tumor protein D54 protein (1-206AA)
- Tumor protein D54 protein (1-206AA)
- Antigen Synonyms:
- 2810411G23Rik|AU016537|C86069|D54|DKFZp686A1765|HCCR binding protein 2|hD54|MGC73020|RP23 33L3.2|TPD52L2|TPD54|TPD54_HUMAN|tumor protein D52 like 2|Tumor protein D52-like 2|Tumor protein D54
- 2810411G23Rik|AU016537|C86069|D54|DKFZp686A1765|HCCR binding protein 2|hD54|MGC73020|RP23 33L3.2|TPD52L2|TPD54|TPD54_HUMAN|tumor protein D52 like 2|Tumor protein D52-like 2|Tumor protein D54
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43399
- O43399
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together