Anti-SLC17A3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC17A3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-774
- 77799-773
- Antigen Symbol:
- SLC17A3
- SLC17A3
- Antigen Name:
- Sodium-dependent phosphate transport protein 4 protein (1-125AA)
- Sodium-dependent phosphate transport protein 4 protein (1-125AA)
- Antigen Synonyms:
- SLC17A3|NPT4|Sodium-dependent phosphate transport protein 4|Na(+)/PI cotransporter 4|Sodium/phosphate cotransporter 4|Solute carrier family 17 member 3
- SLC17A3|NPT4|Sodium-dependent phosphate transport protein 4|Na(+)/PI cotransporter 4|Sodium/phosphate cotransporter 4|Solute carrier family 17 member 3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00476
- O00476
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together