Anti-PIF1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PIF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-193
- 77836-192
- Antigen Symbol:
- PIF1
- PIF1
- Antigen Name:
- ATP-dependent DNA helicase PIF1 protein (125-360AA)
- ATP-dependent DNA helicase PIF1 protein (125-360AA)
- Antigen Synonyms:
- ATP dependent DNA helicase ATP-dependent DNA helicase C15orf20|DNA helicase homolog DNA helicase DNA repair and recombination helicase Petite integration frequency 1|PIF|PIF1 5' to 3' DNA helicase|PIF1 5' to 3' DNA helicase homolog|PIF1 DNA helicase|PIF1/RRM3 DNA helicase like protein|PIF1/RRM3 DNA helicase-like protein|PIF1_HUMAN
- ATP dependent DNA helicase ATP-dependent DNA helicase C15orf20|DNA helicase homolog DNA helicase DNA repair and recombination helicase Petite integration frequency 1|PIF|PIF1 5' to 3' DNA helicase|PIF1 5' to 3' DNA helicase homolog|PIF1 DNA helicase|PIF1/RRM3 DNA helicase like protein|PIF1/RRM3 DNA helicase-like protein|PIF1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H611
- Q9H611
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together