Anti-SOCS1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SOCS1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77800-577
- 77800-578
- Antigen Symbol:
- SOCS1
- SOCS1
- Antigen Name:
- Suppressor of cytokine signaling 1 protein (1-211AA)
- Suppressor of cytokine signaling 1 protein (1-211AA)
- Antigen Synonyms:
- CISH 1|CISH1|Cytokine inducible SH2 protein 1|JAB|JAK binding protein|JAK-binding protein|Janus kinase binding protein|SOCS 1|SOCS-1|SOCS1_HUMAN|SSI 1|SSI-1|SSI1|STAT induced STAT inhibitor 1|STAT-induced STAT inhibitor 1|Suppressor of cytokine signaling 1|Supressor of cytokine signalling 1|TEC interacting protein 3|Tec-interacting protein 3|TIP 3|TIP-3|TIP3
- CISH 1|CISH1|Cytokine inducible SH2 protein 1|JAB|JAK binding protein|JAK-binding protein|Janus kinase binding protein|SOCS 1|SOCS-1|SOCS1_HUMAN|SSI 1|SSI-1|SSI1|STAT induced STAT inhibitor 1|STAT-induced STAT inhibitor 1|Suppressor of cytokine signaling 1|Supressor of cytokine signalling 1|TEC interacting protein 3|Tec-interacting protein 3|TIP 3|TIP-3|TIP3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15524
- O15524
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together