To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Synthesized peptide derived from Histone H2A around the non-phosphorylation site of T121.
- Antigen Symbol:HIST1H2AG/HIST1H2AB/HIST1H2AD/HIST1H2AH/HIST2H2AA3/HIST3H2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P0C0S8/P04908/P20671/Q96KK5/Q6FI13/Q7L7L0
- Antigen Synonyms:HIST1H2AG|H2AFP|HIST1H2AI|H2AFC|HIST1H2AK|H2AFD|HIST1H2AL|H2AFI|HIST1H2AM|H2AFN|Histone H2A type 1|H2A.1|Histone H2A/p|HIST1H2AB|H2AFM|HIST1H2AE|H2AFA|Histone H2A type 1-B/E|Histone H2A.2|Histone H2A/a|Histone H2A/m|HIS
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:5000
Type:
Antigen: HIST1H2AG/HIST1H2AB/HIST1H2AD/HIST1H2AH/HIST2H2AA3/HIST3H2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat