Order Entry
United States
ContactUsLinkComponent
Dr. Ehrenstorfer Difenoconazole D6 (1,1,2,3,3,3-propyl D6) 100 µg/ml in Acetonitrile
Dr. Ehrenstorfer Difenoconazole D6 (1,1,2,3,3,3-propyl D6) 100 µg/ml in Acetonitrile
  77862-650
New Product
 :  
undefined
Dr. Ehrenstorfer Difenoconazole D6 (1,1,2,3,3,3-propyl D6) 100 µg/ml in Acetonitrile
  77862-650
 :  DRE-XA12609010AL
New Product
 :  

 

  • Description:
    Difenoconazole D6 (1,1,2,3,3,3-propyl D6) 100 µg/ml in acetonitrile
  • Size:
    1 ml