Order Entry
United States
ContactUsLinkComponent
Paragon Scientific Distillation, Unleaded Gasoline (Nominal: 37.4 to 181.3 °C)
Paragon Scientific Distillation, Unleaded Gasoline (Nominal: 37.4 to 181.3 °C)
Catalog # 77860-277
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Paragon Scientific Distillation, Unleaded Gasoline (Nominal: 37.4 to 181.3 °C)
Catalog # 77860-277
Supplier:  LGC Standards
Supplier Number:  ALK-CRM-DIGA
New Product
CAS Number:  

Specifications

  • Description:
    Distillation, unleaded gasoline (Nominal: 37.4 to 181.3 °C)
  • Size:
    250 ml

Specifications

About this item