Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Dr. Ehrenstorfer PAH Mix 61 100 to 2000 µg/ml in Acetone/Methanol
Dr. Ehrenstorfer PAH Mix 61 100 to 2000 µg/ml in Acetone/Methanol
Dr. Ehrenstorfer PAH Mix 61 100 to 2000 µg/ml in Acetone/Methanol
Catalog # 77865-006
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Dr. Ehrenstorfer PAH Mix 61 100 to 2000 µg/ml in Acetone/Methanol
Catalog # 77865-006
Supplier:
LGC Standards
Supplier Number:
DRE-XA06100100AM
New Product
CAS Number:
Specifications
Description:
PAH Mix 61 100-2000 µg/ml in acetone/methanol
Size:
1 ml
Specifications
About this item
Product Details
Product Details