Dr. Ehrenstorfer cis-/trans-1,2-Dichloroethene
Catalog # 77861-967
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:540-59-0
- Description:cis-/trans-1,2-dichloroethene
- Size:1 ml
- Formula:C₂H₂Cl₂
- Molecular Weight:96.94 g/mol