CDN Isotopes Histamine-alpha,alpha,beta,beta-d4 2HCl
New Product
- CAS No.:344299-48-5
Frequently Bought Together
J.T.Baker®, Syringe Filters
: Avantor
Acrodisc® Syringe Filters, 25 mm
: Cytiva
VWR® Syringe Filters
: VWR International
SYRINGE FILTER PVDF 0.45UM 30MM NS CS100
: CELLTREAT SCIENTIFIC PRODUCTS MS
Diamond® PureFlow™ MCE Syringe Filters, Sterile, Globe Scientific
: Globe Scientific