Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
CDN-D-3980
CDN-D-3980
Catalog # 77863-562
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
CDN-D-3980
Catalog # 77863-562
Supplier:
LGC Standards
Supplier Number:
CDN-D-3980-0.25G
New Product
CAS Number:
Specifications
CAS No.:
284665-22-1
Description:
1,4-Butane-d8-diamine 2HCl
Size:
250 mg
Specifications
About this item
Product Details
Product Details