Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
CDN Isotopes Ciprofloxacin-d8 HCl H₂O (piperazine-d8)
CDN Isotopes Ciprofloxacin-d8 HCl H₂O (piperazine-d8)
CDN Isotopes Ciprofloxacin-d8 HCl H₂O (piperazine-d8)
Catalog # 77861-096
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
CDN Isotopes Ciprofloxacin-d8 HCl H₂O (piperazine-d8)
Catalog # 77861-096
Supplier:
LGC Standards
Supplier Number:
CDN-D-6729-0.01G
New Product
CAS Number:
Specifications
CAS No.:
1216659-54-9
Description:
Ciprofloxacin-d8 HCl H₂O (piperazine-d8)
Size:
10 mg
Specifications
About this item
Product Details
Product Details