Dr. Ehrenstorfer Dibenz[a,h] Anthracene D14 10 µg/ml in Cyclohexane
Catalog # 77862-556
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:13250-98-1
- Description:Dibenz[a,h] anthracene D14 10 µg/ml in cyclohexane
- Size:10 ml
- Formula:C₂₂D₁₄
- Molecular Weight:292.43 g/mol