CDN Isotopes Buspirone-d8 HCl (n-butyl-d8)
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:1216761-39-5
Specifications
Frequently Bought Together
VWR® Syringe Filters
Supplier: VWR International
Whatman™ Polydisc GW In-Line Filters, Whatman products (Cytiva)
Supplier: Cytiva
Whatman™ Uniflo™ Syringe Filters, Whatman products (Cytiva)
Supplier: Cytiva
J.T.Baker®, Syringe Filters
Supplier: Avantor
Diamond® PureFlow™ CA Syringe Filters, Sterile, Globe Scientific
Supplier: Globe Scientific
Acrodisc® Syringe Filters, 13 mm, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Claristep® Syringeless Filters, Sartorius
Supplier: Sartorius
Acrodisc™ PSF Non-Sterile RC Syringe Filters
Supplier: Cytiva
EZFlow® Syringe Filter, Sample Prep, Hydrophobic PTFE, Foxx Life Sciences
Supplier: Foxx Life Sciences
Target2™ Syringe Filters, Cellulose Acetate
Supplier: Thermo Scientific