Dr. Ehrenstorfer cis-/trans-1,3-Dichloropropene
Catalog # 77861-979
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:542-75-6
- Description:cis-/trans-1,3-dichloropropene
- Size:250 mg
- Formula:C₃H₄Cl₂
- Molecular Weight:110.97 g/mol