0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Standards
Organic Standards
CDN Isotopes n-Dodecylphosphocholine-d38
CDN Isotopes n-Dodecylphosphocholine-d38
CDN Isotopes n-Dodecylphosphocholine-d38
Catalog # 77863-549
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
CDN Isotopes n-Dodecylphosphocholine-d38
Catalog # 77863-549
Supplier:
LGC Standards
Supplier Number:
CDN-D-3612-0.1G
New Product
CAS Number:
Specifications
CAS No.:
130890-78-7
Description:
n-Dodecylphosphocholine-d38
Size:
100 mg
Specifications
About this item
Product Details
Product Details