CDN Isotopes L-Alanine-2,3,3,3-d4
New Product
- CAS No.:18806-29-6
Frequently Bought Together
EZBio® Vent Filter, 0.22 µm, Nonsterile, Foxx Life Sciences
: Foxx Life Sciences
VWR® Gas Venting Filters
: VWR International
CellPro™ Syringe Filters
: Alkali Scientific
LabExact® Trupor® Nylon High Flow, Low Back Pressure Filter Membranes
: I.W. TREMONT CO., INC.
SYRINGE FILTER PTFE 0.22UM 13MM S CS75
: CELLTREAT SCIENTIFIC PRODUCTS MS
FILTER SYS 1000ML PES 0.22UM 90MM S CS12
: VistaLab
Titan3™ PVDF Syringe Filters, Thermo Scientific
: Thermo Scientific
Minisart® RC Syringe Filters
: Sartorius
Syringe Filters, Argos Technologies
: Antyila Scientific
BOT TP FILT PP 1000ML NYLN .45UM NS CS12
: VistaLab
LabExact® Polypropylene Syringe Filters
: I.W. TREMONT CO., INC.