Order Entry
United States
ContactUsLinkComponent
Mikromol 8-Chloro-11-(piperazin-1-yl)-5H-dibenzo[b,e][1,4]diazepine
Mikromol 8-Chloro-11-(piperazin-1-yl)-5H-dibenzo[b,e][1,4]diazepine
Catalog # 77862-957
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Mikromol 8-Chloro-11-(piperazin-1-yl)-5H-dibenzo[b,e][1,4]diazepine
Catalog # 77862-957
Supplier:  LGC Standards
Supplier Number:  MM0480.03
New Product
CAS Number:  

Specifications

  • CAS No.:
    6104-71-8
  • Description:
    8-Chloro-11-(piperazin-1-yl)-5H-dibenzo[b,e][1,4]diazepine
  • Size:
    100 mg
  • Formula:
    C₁₇H₁₇ClN₄
  • Molecular Weight:
    312.8 g/mol

Specifications

About this item