Dr. Ehrenstorfer Triphenyl phosphate (TPP) 500 µg/ml in Methyl-tert-butyl ether
Catalog # 77864-906
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:115-86-6
- Description:Triphenyl phosphate (TPP) 500 µg/ml in Methyl-tert-butyl ether
- Size:1 ml
- Formula:C₁₈H₁₅O₄P
- Molecular Weight:326.29 g/mol