Dr. Ehrenstorfer Fumonisin B2 50 μg/ml in Acetonitrile/Water
Catalog # 77862-584
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:116355-84-1
- Description:Fumonisin B2 50 μg/ml in Acetonitrile/Water
- Size:5 ml
- Formula:C₃₄H₅₉NO₁₄
- Molecular Weight:705.839 g/mol