$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
Standards
Organic Standards
Dr. Ehrenstorfer Acenaphthene D10
Dr. Ehrenstorfer Acenaphthene D10
77862-359
:
LGC Standards
New Product
:
Dr. Ehrenstorfer Acenaphthene D10
77862-359
:
LGC Standards
:
DRE-C20505100
New Product
:
CAS No.:
15067-26-2
Description:
Acenaphthene D10
Size:
100 mg
Formula:
C₁₂D₁₀
Molecular Weight:
164.13 g/mol