Lipomed Ethyl-Beta-D-glucuronide
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:17685-04-0
- Formula:C₈H₁₄O₇
- Molecular Weight:222.19 g/mol
Specifications
Frequently Bought Together
Whatman™ Puradisc Syringe Filters, CA, Whatman products (Cytiva)
Supplier: Cytiva
Acrodisc® Syringe Filters, 13 mm, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Whatman™ Uniflo™ Syringe Filters, Whatman products (Cytiva)
Supplier: Cytiva
VWR® Vacuum Filtration Systems, Standard Line
Supplier: VWR International
Acrodisc® Syringe Filters for Prefiltration, Cytiva
Supplier: Cytiva
J.T.Baker®, Syringe Filters
Supplier: Avantor
Diamond® PureFlow™ PTFE Syringe Filters, Sterile, Globe Scientific
Supplier: Globe Scientific
Target® Syringe Filters, PVDF, Thermo Scientific
Supplier: Thermo Scientific
Diamond® PureFlow™ PVDF Sryinge Filters, Sterile, Globe Scientific
Supplier: Globe Scientific
FILTER SYS 500ML PES 0.22UM 90MM S CS12
Supplier: CELLTREAT SCIENTIFIC PRODUCTS MS
Syringe Filters, Color Coded, MicroSolv Technology
Supplier: MicroSolv Technology Corp
FILTER SYS 500ML PES 0.45UM 90MM S CS12
Supplier: VistaLab