Thioanisole ≥99.0%
About this item
CAS Number: 100-68-5 MDL Number: MFCD00008559 Molecular Formula: C7H8S Molecular Weight: 124.20 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 188 Flash Point (°C): 57 Specific Gravity (20/20): 1.06
Product Categories: Synthetic Organic Chemistry, Peptide Synthesis Other Product Names / Synonyms: Methyl Phenyl Sulfide, Methylthiobenzene
- CAS Number:100-68-5
- Formula:C₇H₈S
- MDL Number:MFCD00008559
- Molecular Weight:124.21 g/mol
- Boiling Pt:188 °C
- Melting Pt:–15 °C
- Density:1.06 (20 °C)
- Flash Pt:57 °C
- Storage Temperature:Ambient
- EINECS:202-878-2
- UN:1993
- ADR:3,III
3 Options Available Below
Product Details & Documents
CAS Number: 100-68-5 MDL Number: MFCD00008559 Molecular Formula: C7H8S Molecular Weight: 124.20 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 188 Flash Point (°C): 57 Specific Gravity (20/20): 1.06
Product Categories: Synthetic Organic Chemistry, Peptide Synthesis Other Product Names / Synonyms: Methyl Phenyl Sulfide, Methylthiobenzene
- CAS Number:100-68-5
- Formula:C₇H₈S
- MDL Number:MFCD00008559
- Molecular Weight:124.21 g/mol
- Boiling Pt:188 °C
- Melting Pt:–15 °C
- Density:1.06 (20 °C)
- Flash Pt:57 °C
- Storage Temperature:Ambient
- EINECS:202-878-2
- UN:1993
- ADR:3,III
Specifications for Product Family
Specification Test Results
- Purity/Analysis Method:99.0% (GC)
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



