Order Entry
United States
Orders LinkContactUsLinkComponent
(2-Methoxyethoxy)methyl chloride ≥95.0%

(2-Methoxyethoxy)methyl chloride ≥95.0%

(2-Methoxyethoxy)methyl chloride ≥95.0%
Supplier:  TCI America
Certificates
(2-Methoxyethoxy)methyl chloride ≥95.0%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

CAS Number: 3970-21-6 MDL Number: MFCD00000888 Molecular Formula: C4H9ClO2 Molecular Weight: 124.56 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Boiling point (°C): 52 Flash Point (°C): 54 Specific Gravity (20/20): 1.10 Storage Temperature: 0-10°C

Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis) Other Product Names / Synonyms: 1-Chloromethoxy-2-methoxyethane, Ethylene Glycol Chloromethyl Methyl Ether, MEM-Chloride

Synonyms:  1-Chloromethoxy-2-methoxyethane, 1-Chloro-2,5-dioxahexane, MEM chloride
  • CAS Number:3970-21-6
  • Formula:ClCH₂OCH₂CH₂OCH₃
  • MDL Number:MFCD00000888
  • Molecular Weight:124.57 g/mol
  • Boiling Pt:52 °C
  • Density:1.10 (20 °C)
  • Flash Pt:54 °C
  • EINECS:223-589-8
  • UN:1992
  • ADR:3,III

Documents

Certificates
Search by Lot Number
Specifications for Product Family

Specification Test Results

  • Purity/Analysis Method:
    95.0% (GC)
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 25 mL
    Log in for availability
    Log in for availability
    Each (25ml)
    Each (25ml) - $78.00
    $78.00
    Each (25ml)
       $78.00

    Supplier Number: M0680-25ML
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 500 mL
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $597.00
    $597.00
    Each (500ml)
       $597.00

    Supplier Number: M0680-500ML

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.