Order Entry
United States
Orders LinkContactUsLinkComponent
Ethyl methacrylate ≥99.0% stabilised

Ethyl methacrylate ≥99.0% stabilised

Flammable
Irritant
Supplier:   TCI America
Ethyl methacrylate ≥99.0% stabilised
Supplier:  TCI America
Certificates
Ethyl methacrylate ≥99.0% stabilised

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

(stabilized with HQ) CAS Number: 97-63-2 MDL Number: MFCD00009161 Molecular Formula: C6H10O2 Molecular Weight: 114.14 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 117 Melting point (°C): -75 Flash Point (°C): 20 Specific Gravity (20/20): 0.91

Product Categories: Materials Chemistry, Monomers Other Product Names / Synonyms: Methacrylic Acid Ethyl Ester

Synonyms:  Ethyl 2-methylpropenoate, Methacrylic acid ethyl ester, Ethyl-2-methylprop-2-enoat, Ethyl-2-methylpropenoat, Ethyl 2-methylprop-2-enoate
  • CAS Number:97-63-2
  • Formula:C₆H₁₀O₂
  • MDL Number:MFCD00009161
  • Molecular Weight:114.14 g/mol
  • Boiling Pt:117 °C
  • Melting Pt:–75 °C
  • Density:0.91 (20 °C)
  • Flash Pt:20 °C
  • EINECS:202-597-5
  • ADR:3,II
  •  :Stabilised with Stabilised
  • UN:2277

Documents

Certificates
Search by Lot Number
Specifications for Product Family

Specification Test Results

  • Purity/Analysis Method:
    99.0% (GC)
Product Family Options

Product Information

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.