Order Entry
United States
ContactUsLinkComponent
Food Green No. 3
Food Green No. 3
Catalog # TCF0146-010G
Supplier:  TCI America
CAS Number:  
Food Green No. 3
Catalog # TCF0146-010G
Supplier:  TCI America
Supplier Number:  F0146-10G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • CAS No.:
    2353-45-9
  • Description:
    Food Green No. 3
  • Size:
    10 g
  • Formula:
    C₃₇H₃₄N₂Na₂O₁₀S₃
  • Description:
    Food Green No. 3
  • Cat. No.:
    TCF0146-010G
  • Supplier no.:
    F0146-10G

Specifications

About this item

Color index 42053

Form: Powder
Color: Green