Order Entry
United States
Orders LinkContactUsLinkComponent
Copper(II) acetate monohydrate ≥95.0%

Copper(II) acetate monohydrate ≥95.0%

Corrosive
Irritant
Environment
Supplier:   TCI America
Copper(II) acetate monohydrate ≥95.0%
Supplier:  TCI America
Certificates
Copper(II) acetate monohydrate ≥95.0%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 6046-93-1 MDL Number: MFCD00149570 Molecular Formula: C4H6CuO4 Molecular Weight: 181.63 Purity/Analysis Method: 95.0% (T) Form: Crystal Melting point (°C): 115

Product Categories: Synthetic Organic Chemistry, Activation Reaction of Unactivated Bonds, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction, Metal Catalysts & Ligands (Click Chemistry), Materials Chemistry, Magnetic Metal Complexes (Magn Other Product Names / Synonyms: Acetic Acid Copper(II) Salt

Synonyms:  Copper diacetate monohydrate, Cupric acetate monohydrate, Acetic acid copper (II) salt monohydrate, Copper(II) acetate hydrate
  • CAS Number:6046-93-1
  • Formula:(H₃CCOO)₂Cu·H₂O
  • MDL Number:MFCD00149570
  • Molecular Weight:199.65 g/mol
  • Merck Index:13,02651
  • Melting Pt:115 °C
  • Density:1.88 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:205-553-3
  • UN:3077
  • ADR:9,III

Documents

Specifications for Product Family

Specification Test Results

  • Purity/Analysis Method:
    95.0% (T)
Product Family Options

Product Information

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.