Spherometer
Spherometer
Catalog # 470137-716
Supplier: United Scientific Supplies
: SPRM01
Jump to:
Product Details & Documents
Precision Measurement of Very Small Lengths
- Easy to read dials
- Accurate to 0.01 mm
This useful instrument determines the radius of curvature of spherical surfaces such as mirrors and lenses. Its micrometer screw is graduated to measure displacement as small as 0.01mm.
Specifications
- Description:Spherometer