



Methylamine 2 M in methanol
Catalog # AAH26889-AE
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Size:100 ml
- Cat. No.:AAH26889-AE
- Supplier no.:H26889-AE
Specifications
About this item
:
:31.06
Merck Index:14,6014
Boiling Pt:ca 40 °C
Boiling Pt:∼ 40 °C
Density:0.785
Flash Pt:7 °C(45 °F)
Flash Pt:7 °C (45 °F)
Storage Temperature:Ambient
EINECS:200-820-0
UN:3286
ADR:"3",II
:GHS02
:GHS05
:GHS06
:GHS08
CAS Number:74-89-5
Formula:CH₅N
MDL Number:MFCD00008104
ADR:3,II