Silver, AAS Standard Solution, Specpure®, Ag 1000 µg/ml, Thermo Scientific
Catalog # AA88096-AE
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Concentration:1000 µg/ml
- Matrix:5% HNO₃
- Size:100 ml
- MDL No.:MFCD00003397
- Cat. No.:AA88096-AE
- Element:Silver
- Supplier no.:88096-AE
Specifications
About this item
Molecular Formula: Ag in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00003397