ContactUsLinkComponent
Ethanol denatured ≥90% for spectrophotometry (denatured with 5% isopropanol and 5% methanol)
Flammable
Health
Ethanol denatured ≥90% for spectrophotometry (denatured with 5% isopropanol and 5% methanol)
Certificates
Flammable
Health
Ethanol denatured ≥90% for spectrophotometry (denatured with 5% isopropanol and 5% methanol)
Certificates
Specification Test Results
  • Evaporation residue:
    5ppm max.
  • H2O:
    0.1% max.