

Ethanol denatured, anhydrous 94-96% ACS (denatured with 5% isopropanol and 5% methanol)
:64-17-5
:
:H₃CCH₂OH
:MFCD00003568
:46.07 g/mol
:14,3760
:78 °C
:–90 °C
:0.789
:11 °C (52 °F)
:Ambient
:200-578-6
:1987
:3,II
:GHS02
:GHS08
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:EPA Safer chemical solvent (green circle rating); Chem21 solvent selection guide recommended by default and after discussion
- Appearance:Pass test.
- Color (APHA):10
- Refractive index:1.3589-1.3599
- Residual water (KF Titration):<0.5%
- Specific gravity (25 °C):0.785 - 0.795
- Residue on Evaporation:0.001%
Frequently Bought Together