Order Entry
United States
Orders LinkContactUsLinkComponent
 

Picolinamide 98%

Picolinamide 98%
Certificates
Picolinamide 98%
Picolinamide 98%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 98% CAS No.: 1452-77-3 Molecular Formula: C6H6N2O Formula Weight: 122.13 Melting Point: 104-108° Boiling Point: 143°/20mm MDL No.: MFCD00023483

Synonyms:  2-Pyridinecarboxamide, Pyridine-2-carboxamide
  • CAS Number:1452-77-3
  • Formula:C₆H₆N₂O
  • MDL Number:MFCD00023483
  • Molecular Weight:122.13 g/mol
  • Boiling Pt:143 °C (20 mmHg)
  • Boiling Pt:143 °C/20 mm
  • Melting Pt:104…108 °C
  • Storage Temperature:Ambient
  • EINECS:215-921-5
Product Family Options

Product Information

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.