Multi-Element Plasma Standard Solution 4, Specpure®, Thermo Scientific
AA42885-AE
:
- Size:100 ml
- MDL No.:MFCD00212340
- Cat. No.:AA42885-AE
- Supplier no.:42885-AE
Molecular Formula: Matrix: dilute HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00212340