Order Entry
United States
Orders LinkContactUsLinkComponent
 

Magnesium nitrate solution 1% in nitric acid 5%, Specpure® matrix modifier solution

100 ml
Catalog # AA39044-AE
Corrosive
 :  039044-AE
Magnesium nitrate solution 1% in nitric acid 5%, Specpure® matrix modifier solution
Catalog # AA39044-AE
CAS Number:  10377-60-3
Magnesium nitrate solution 1% in nitric acid 5%, Specpure® matrix modifier solution

100 ml

CAS Number:  10377-60-3
Price
$124.74
Log in for availability
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Molecular Formula: 1% Mg(NO3)2 in 2% HNO3 Physical Form: Liquid UN No.: 3264 MDL No.: MFCD00011103

  • CAS Number:10377-60-3
  • Formula:Mg(NO₃)₂
  • MDL Number:MFCD00011103
  • Storage Temperature:Ambient
  • EINECS:231-714-2
  • UN:3264
  • ADR:8,III
Specifications

Specifications

  • Size:
    100 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.