Order Entry
United States
Orders LinkContactUsLinkComponent
Iron(III) hydroxide, γ-phase

Iron(III) hydroxide, γ-phase

Iron(III) hydroxide, γ-phase
Certificates
Iron(III) hydroxide, γ-phase

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS No.: 20344-49-4 Molecular Formula: FeO(OH) Formula Weight: 88.85 Physical Form: -60 Mesh Powder Odor: Odorless Melting Point: 135° dec. MDL No.: MFCD00064782

  • CAS Number:20344-49-4
  • Formula:FeO(OH)
  • MDL Number:MFCD00064782
  • Molecular Weight:88.85 g/mol
  • Merck Index:14,4025
  • Melting Pt:135 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:243-746-4

Documents

Product Family Options

Product Information

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.