Silver two part conductive adhesive, EXP. XX/XX resin and hardener
Supplier: Thermo Scientific Chemicals
Certificates
About this item
:
MDL Number:MFCD00003397
Storage Temperature:Ambient
Related Information
Certificates
Search by Lot NumberAlfa Aesar Metals and Materials
Product Literature
Alfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
Frequently Bought Together